To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:B220
- Clonality:Monoclonal
- Clone:MEM-56
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Antigen symbol:CD45RA
- Conjugation:PerCP-Cyanine 5.5
- Flow cytometry:Yes
- Size:100 tests
- Form:Liquid
- Antigen synonyms:CD45R|CD45|CD45RA|GP180|Leukocyte common antigen|L-CA|LCA|CD45 antigen|EC 3.1.3.48
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Human thymocytes and T lymphocytes
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 5.5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Cat. no.:76965-562
- Packaging:Glass bottle
Specifications
About this item
Mouse monoclonal [MEM-56] antibody to CD45RA (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from Human.
- Validated Applications: Flow Cytometry
- Recommended Dilutions: Flow Cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: MEM-56
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human