To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:A26C1A
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Monkey,Human,Dog,Rat,Mouse
- Antigen symbol:beta Actin
- Conjugation:Unconjugated
- ImmunoFluorescence:Yes
- Size:600 µg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Actin, cytoplasmic 1, N-terminally processed|b actin|ACTB|A26C1B|Actx|Actin beta|ACTB_HUMAN|Actin cytoplasmic 1|beta Actin
- Storage buffer:Supplied in Phosphate Buffered Saline with 20% Glycerol and 0.05% Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
- Concentration:3 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant peptide derived from within residues a.a. 100 to the N terminus of human beta Actin, expressed in and purified from E. coli
- Purification:Affinity purification
- Cat. no.:76836-586
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to beta Actin for WB and IF with samples derived from Human, Rat, Mouse, Canine and Monkey.
- Validated Applications: WB, IF
- Recommended Dilutions: WB: 1:250 to 1:2,000, IF: 1 to 50 to 1:250
Type: Primary
Antigen: beta Actin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat, Mouse, Dog, Monkey