Anti-Random Protein Goat Polyclonal Antibody
76834-842
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Antigen symbol:Random Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Size:600 µg
- Environmentally Preferable:
- Form:Liquid
- Storage buffer:Supplied in Phosphate Buffered Saline with 20% Glycerol and 0.05% Sodium Azide
- Sequence:ALRRTARNPRSCGRRHRGRWRRCADPCTHRDRRSDRRERRPERRHQDRAARDGRAAAPDRHERWRRSERRGGGSCSRQGQLRLQRSNGRV
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at 4 °C. Avoid freeze/thaw cycles.
- Concentration:3 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C.
- Immunogen:Recombinant random protein fragment, expressed in and purified from E. coli
- Purification:Affinity purification
- Cat. no.:76834-842
- Packaging:Plastic vial
Goat polyclonal antibody to Random Protein for WB.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
- Host: Goat
- Clonality: Polyclonal
- Isotype: IgG
- Conjugate: Unconjugated
Type: Primary
Antigen: Random Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: