About this item
Mouse monoclonal (ABT-CD45) antibody to CD45 for IHC with samples derived from Human.
- Validated Applications: IHC
Type: Primary
Antigen: CD45RO
Clonality: Monoclonal
Clone: ABT-CD45
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 77073-598
- 77072-372
- Antigen name:
- B220
- B220
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- ABT-CD45
- ABT-CD45
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P08575
- UniprotID# P08575
- Isotype:
- IgG
- IgG