Anti-Neuronal calcium-binding protein Rabbit Polyclonal Antibody
Catalog # 77056-886
Supplier: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:amyloid beta (A4) precursor protein binding, family A, member 2 binding protein
- Antigen symbol:Neuronal calcium-binding protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q96P71
- Antigen synonyms:Amyloid beta A4 protein-binding family A member 2-binding protein|dJ63M2.5|N-terminal EF-hand calcium-binding protein 3|N terminal EF hand calcium binding protein 3|EFCBP3|APBA2BP|NECA3_HUMAN|Amyloid beta A4 protein binding family A member 2 binding protein|dJ63M2.4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:44 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human NECAB3 (amino acids 321-370)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77056-886
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to NECAB3 for WB, IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: Neuronal calcium-binding protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse