Anti-TUBGCP5 Rabbit Polyclonal Antibody
Catalog # 77054-630
Supplier: ANTIBODIES.COM LLC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Gamma tubulin complex component GCP5
- Antigen symbol:TUBGCP5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:UniprotID# Q96RT8
- Antigen synonyms:GCP 5|Tubulin gamma complex associated protein 5|OTTHUMP00000159254|KIAA1899|GCP5_HUMAN|GCP-5|Gamma-tubulin complex component 5|TUBGCP5
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular weight:118 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TUBGCP5 (amino acids 741-790)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. no.:77054-630
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to TUBGCP5 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: TUBGCP5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat