Order Entry
Canada
ContactUsLinkComponent
Mouse Neuropilin 1 ELISA Kit
Mouse Neuropilin 1 ELISA Kit
Catalog # 77207-406
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Neuropilin 1 ELISA Kit
Catalog # 77207-406
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A311016-96
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Host:
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Neuropilin 1
  • Description:
    Mouse Neuropilin 1 ELISA kit
  • Size:
    96 tests
  • Environmentally Preferable:
  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    0.5 - 8 ng/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    0.023 ng/ml
  • Regulatory status:
    RUO
  • Cat. no.:
    77207-406

Specifications

About this item

Mouse Neuropilin 1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Neuropilin 1 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 0.5 to 8 ng/ml
  • Sensitivity: 0.023 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse Neuropilin 1 ELISA kit (A311016) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Neuropilin 1 in serum, plasma or other biological fluids. An antibody specific for Neuropilin 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Neuropilin 1 present in each sample is bound to the wells by the immobilized antibody. Biotinylated Anti-Neuropilin 1 antibody, which also binds the Neuropilin 1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Neuropilin 1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Neuropilin 1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Neuropilin 1 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Neuropilin 1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.