Order Entry
Canada
ContactUsLinkComponent
Human Golgin-245 ELISA Kit
Human Golgin-245 ELISA Kit
  77175-546
 :  
undefined
Human Golgin-245 ELISA Kit
  77175-546
 :  A314315-96
 :  

 

  • Sample type:
    Serum, plasma or other biological fluids
  • Detection method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    100 - 1600 ng/L
  • Storage temperature:
    4 °C
  • Sample volume:
    40 μl
  • Sensitivity:
    6.79 ng/L
  • Regulatory status:
    RUO
  • Cat. no.:
    77175-546

 

 

Human Golgin-245 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Golgin-245 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 100 - 1600 ng/L
  • Sensitivity: 6.79 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human Golgin-245 ELISA kit (A314315) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Golgin-245 in serum, plasma or other biological fluids. An antibody specific for Golgin-245 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Golgin-245 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-Golgin-245 antibody, which also binds the Golgin-245 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Golgin-245 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Golgin-245 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Golgin-245 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Golgin-245 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.