Snap Cap
Catalog #: 76808-352
Supplier: THOMSON INSTRUMENT CO
Specifications
- Cap Size:11 mm
- Sterility:Non sterile
- Color:Clear
- Description:Snap cap
- Material:PTFE/rubber septa
- Cat. no.:76808-352
Specifications
About this item
Customers Who Bought This Also Bought
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
$176.89 to $1,730.69
N,N-Dimethylformamide ≥99.8% (by GC), GR ACS, Supelco®
Supplier: MilliporeSigma
$1,286.88 to $4,894.43
EZFlip Conical Centrifuge Tubes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$94.31 to $107.08
CrossLab Autosampler Probes, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
$676.03
Nunc-Immuno™ and Fluoronunc™/Luminunc™ 96-Well Modules and Frames, Thermo Scientific
Supplier: Thermo Fisher Scientific
$1,194.01
D-(+)-Sucrose, crystals NF, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
$447.91 to $1,264.81
Pentane ≥98.0%, Laboratory Reagent, VWR Chemicals BDH®
Supplier: VWR International
$500.19 to $1,790.36
Muscovite Mica Sheets, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
$69.52 to $88.34