To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Unknown compositions - ferrous ammonium sulfate
- Cat. no.:470352-538
- Includes:Instructions and SDS
- No. of Uses:25
Specifications
About this item
Determine the purity of a ferrous ammonium sulfate sample via gravimetric and spectrophotometric analysis.
- Contains enough materials needed for 25 groups
- Teachers Manual and Student Study Guide copymasters are included
In this AP Chemistry experiment, students will perform Gravimetric Analysis and Spectrophotometric analysis to determine the purity of ferrous ammonium sulfate samples of unknown composition.
5 Lettered Impure Ferrous Ammonium Sulfate Samples, 25 g
3×50 ml Ammonia Solution
Phenanthroline, 12.5 g