To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Unknown compositions - potassium hydrogen phthalate
- Cat. no.:470352-536
- Includes:Instructions and SDS
- No. of Uses:25
Specifications
About this item
Determine the purity of a potassium hydrogen phthalate sample via acid-base titration.
- Contains enough materials needed for 25 groups
- Teachers Manual and Student Study Guide copymasters are included
Learn the importance of substance purity at an AP Chemistry level by determining the composition of a set of impure potassium hydrogen phthalate samples via the one of the most popular analytical methods: Acid-Base Titration.
Sodium Hydroxide Bead, 125 g
2×500 ml Hydrochloric Acid Solution, 1 M
2×30 ml Bromothymol Blue Indicator