To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Method / Format:Column (silica-based)
- Prep size:Giga
- Refine target molecule:Plasmid (endotoxin-free)
- Sample type:Bacteria
- Target molecule:Plasmid
- Description:EndoZero III spin-column assemblies
- Size:5 pack
- Cat. no.:77020-280
Specifications
About this item
The EndoZero III spin-column can be used with centrifuges for the removal of residual endotoxins in plasmid DNA.
- Isolate up to 25 mg of endotoxin-free plasmid DNA from E. coli
Available as a refill for the ZymoPURE™ II plasmid gigaprep kit. The EndoZero III spin-column with 15 ml reservoir-X features durable polypropylene construction and can be used with centrifuges for the removal of residual endotoxins in plasmid DNA prepared with the ZymoPURE™ II plasmid gigaprep kit, which is the fastest and simplest method available. Capacity with 15 ml Reservoir-X is 15 ml.