Anti-SLURP1 Rabbit Polyclonal Antibody
Catalog # 76764-132
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Secreted LY6/PLAUR domain containing 1
- Antigen symbol:SLURP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:50 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:57152
- Antigen synonyms:MDM|secreted Ly-6/uPAR-related protein 1|lymphocyte antigen 6-like secreted|secreted Ly6/uPAR related protein 1|SLURP-1|LY6LS|ARS|anti-neoplastic urinary protein|ARS(component B)-81/S|ANUP|LY6-MT|ArsB
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-103 of human SLURP1 (NP_065160.1).
- Purification:Affinity purification
- Cat. no.:76764-132
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SLURP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse