Anti-DHRS9 Rabbit Polyclonal Antibody
Catalog # 76749-788
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Dehydrogenase/reductase 9
- Antigen symbol:DHRS9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:10170
- Antigen synonyms:SDR9C4|retinol dehydrogenase 15|dehydrogenase/reductase SDR family member 9|short chain dehydrogenase/reductase family 9C member 4|dehydrogenase/reductase (SDR family) member 9|RDHTBE|RDHL|tracheobronchial epithelial cell-specific retinol dehydrogenase|RDH15|RDH-E2|NADP-dependent retinol dehydrogenase/reductase|RDH-TBE|3ALPHA-HSD|3-alpha hydroxysteroid dehydrogenase|retinol dehydrogenase homolog|short-chain dehydrogenase/reductase retSDR8|RETSDR8|3-alpha-HSD
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 18-319 of human DHRS9 (NP_001135743.1).
- Purification:Affinity purification
- Cat. no.:76749-788
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: DHRS9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat