Anti-GRIN2B Rabbit Polyclonal Antibody
Catalog #: 76757-972
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
- Clonality:Polyclonal
- Gene ID:2904
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Antigen symbol:GRIN2B
- Conjugation:Unconjugated
- ImmunoFluorescence:Yes
- Size:50 µl
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:glutamate receptor|hNR3|GRIN2B|ionotropic|N-methyl D-aspartate 2B|NR2B|N-methyl-D-aspartate receptor subunit 2B|HGNC:4586|NMDAR2B
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:Blue ice
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 1400 to the C-terminus of human GRIN2B (NP_000825.2).
- Purification:Affinity purification
- Cat. no.:76757-972
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Customers Who Bought This Also Bought