Cynomolgus Monkey Recombinant M-CSF R Protein (from HEK293 Cells)
Catalog #: 77131-370
Supplier: Prosci
New Product
CAS Number:
Specifications
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:CD115|CSFR|M-CSFR|C-FMS|FMS|FIM2|CSF1R
- Protein/peptide name:M-CSF R
- Purity:>95% as determined by SDS-PAGE. Endotoxin level is less than 1.0 EU per µg by the LAL method.
- Molecular weight:57.1 kDa
- Sequence:Ile 99 - Pro 596
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping temperature:Room temperature
- Tested applications:WB
Specifications
About this item
Customers Who Bought This Also Bought