BTLA Peptide
Catalog #: 77122-374
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:151888
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Mouse
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:B- and T-lymphocyte attenuator|B- and T-lymphocyte-associated protein|BTLA1|CD272
- Protein/peptide name:BTLA
- Formulation:Lyophilized. BTLA peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought