Pirin Peptide
Catalog #: 77132-422
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:8544
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:Probable quercetin 2|Pirin|3-dioxygenase PIR|Probable quercetinase
- Protein/peptide name:PIR
- Formulation:Lyophilized. Pirin peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought