Anti-PF-03446962 Recombinant Antibody
Catalog #: 76763-100
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Isotype:IgG1 kappa
- Conjugation:Unconjugated
- Size:0.1 mg
- Antigen synonyms:hPAM4-DOTA|hPAM4|hPAM4 IgG-DOTA
- Modification:None
- Storage buffer:PBS buffer pH 7.5 .
- Storage temperature:Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 ...730 °C for 1-2 weeks. Store at –20…+730 °C for 12 months. Store at –80…+730 °C for long term storage.
- Concentration:Batch dependent
- Shipping temperature:Blue ice
- Immunogen:Humanized/MUC1/CD227 [Homo sapiens].
- Purification:>95%
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen:
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1-kappa
Reactivity:
Customers Who Bought This Also Bought