To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:ring finger protein 26
- Clonality:Polyclonal
- Gene ID:79102
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Antigen symbol:RNF26
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.1 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:HM89|RNF26 Antibody: FB22|WHIM|NPYR|NPYRL|D2S201E|NPY3R|NPYY3R|LCR1|HSY3RR|CD184|LAP3|LESTR
- Modification:None
- Storage buffer:RNF26 Antibody is supplied in PBS containing 0.02% sodium azide.
- Concentration:1 mg/ml
- Shipping temperature:Blue ice
- Immunogen:RNF26 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human RNF26.
- Purification:RNF26 Antibody is affinity chromatography purified via peptide column.
- Packaging:Vial
Specifications
About this item
RNF26 antibody can be stored at 4 to 730 °C for three months and –20 to +730 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Type: Primary
Antigen: RNF26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat