MAP3K7IP3 Peptide
Catalog #: 77127-802
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:257397
- Packaging:Vial
- Protein/peptide type:Synthetic
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:MAP3K7IP3|NAP1|Mitogen-activated protein kinase kinase kinase 7-interacting protein 3|TGF-beta-activated kinase 1 and MAP3K7-binding protein 3|TAB-3
- Protein/peptide name:TAB3
- Formulation:Lyophilized. MAP3K7IP3 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77127-802
Specifications
About this item
Customers Who Bought This Also Bought