FOXN2 Peptide
Catalog #: 77128-594
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:3344
- Packaging:Vial
- Protein/peptide type:Synthetic
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:Forkhead box protein N2|HTLF|Human T-cell leukemia virus enhancer factor
- Protein/peptide name:FOXN2
- Formulation:Lyophilized. FOXN2 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought