ATF7 Peptide
Catalog #: 77132-004
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:11016
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:ATFA|Cyclic AMP-dependent transcription factor ATF-7|Activating transcription factor 7|cAMP-dependent transcription factor ATF-7
- Protein/peptide name:ATF7
- Formulation:Lyophilized. ATF7 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77132-004
Specifications
About this item
Customers Who Bought This Also Bought