CDKN2A Peptide
Catalog #: 77120-580
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:1029
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:CDKN2|MLM|INK4|MTS-1|P16|P16INK4|P19|P16-INK4A|P16INK4A|INK4A|Cyclin-dependent kinase 4 inhibitor A|P19ARF|CDK4I|CMM2|P14ARF|P14|ARF|MTS1|TP16
- Protein/peptide name:CDKN2A
- Formulation:Lyophilized. CDKN2A peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought