EXOG Peptide
Catalog #: 77129-264
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:9941
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:endonuclease G-like 1|ENGLA|ENDOGL1|ENGL-b|mitochondrial|ENGLB|endonuclease G-like|ENGL|nuclease EXOG|endo G-like 1|endo/exonuclease (5'-3')|EXOG|ENGL-a|endonuclease G-like 2|ENDOGL2
- Protein/peptide name:EXOG
- Formulation:Lyophilized. EXOG peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77129-264
Specifications
About this item
Customers Who Bought This Also Bought