CD13/ANPEP Peptide
Catalog #: 77120-754
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:290
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:ANPEP|AP-N|PEPN|aminopeptidase M|GP150|aminopeptidase N|CD13|microsomal aminopeptidase|APN|alanyl (membrane) aminopeptidase|myeloid plasma membrane glycoprotein CD13|hAPN|LAP1|alanyl aminopeptidase|P150|AP-M
- Protein/peptide name:CD13/ANPEP
- Formulation:Lyophilized. CD13/ANPEP peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought