neuron navigator 2 Peptide
Catalog #: 77132-456
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:89797
- Packaging:Vial
- Protein/peptide type:Synthetic
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:retinoic acid inducible gene in neuroblastoma 1|APC down-regulated 1|HELAD1|pore membrane and/or filament-interacting-like protein 2|POMFIL2|UNC53H2|NAV2|helicase|unc-53 homolog 2|neuron navigator 2|steerin-2|RAINB1|STEERIN2
- Protein/peptide name:Neuron navigator 2
- Formulation:Lyophilized. neuron navigator 2 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77132-456
Specifications
About this item
Customers Who Bought This Also Bought