filamin A Peptide
Catalog #: 77125-090
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:2316
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:endothelial actin-binding protein|OPD1|OPD2|alpha|XLVD|filamin A|FMD|OPD|CVD1|XMVD|alpha-filamin|FLN-A|FLN|non-muscle filamin|FLN1|ABP-280|FLNA|actin binding protein 280|filamin-1|NHBP|MNS|ABPX|filamin-A
- Protein/peptide name:filamin A
- Formulation:Lyophilized. filamin A peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77125-090
Specifications
About this item
Customers Who Bought This Also Bought