HCoV-OC43 Recombinant Human Coronavirus Nucleocapsid OC43 Protein (from bacteria)
Catalog #: 77126-560
Supplier: Prosci
New Product
CAS Number:
Specifications
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:Bacteria
- Species:HCoV-OC43
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:Human CoV OC43 Nucleocapsid Protein|Human CoV OC43 N Protein
- Protein/peptide name:Nucleocapsid
- Purity:GSH-Agarose (0.65)
- Molecular weight:Mono-Isotopic Mass: 76, 066.25 daltons Average Mass: 76, 113.93 daltons
- Sequence:MSPILGYWKIKGLVQPTRLLLEYLEEKYEEHLYERDEGDKWRNKKFELGLEFPNLPYYIDGDVKLTQSMAIIRYIADKHNMLGGCPKERAEISMLEGAVLDIRYGVSRIAYSKDFETLKVDFLSKLPEMLKMFEDRLCHKTYLNGDHVTHPDFMLYDALDVVLYMDPMCLDAFPKLVCFKKRIEAIPQIDKYLKSSKYIAWPLQGWQATFGGGDHPPKSDLEVLFQGPLGSMSFTPGKQSSSRASSGNRSGNGILKWADQSDQFRNVQTRGRRAQPKQTATSQQPSGGNVVPYYSWFSGITQFQKGKEFEFAEGQGVPIAPGVPATEAKGYWYRHNRRSFKTADGNQRQLLPRWYFYYLGTGPHAKDQYGTDINGVYWVASNQADVNTPADIVDRDPSSDEAIPTRFPPGTVLPQGYYIEGSGRSAPNSRSTSRTSSRASSAGSRSRANSGNRTPTSGVTPDMADQIASLVLAKLGKDATKPQQVTKHTAKEVRQKILNKPRQKRSPNKQCTVQQCFGKRGPNQNFGGGEMLKLGTSDPQFPILAELAPTAGAFFFGSKLELAKVQNLSGNPDEPQKDVYELRYNGAIRFDSTLSGFETIMKVLSENLNAYQQQDGMVNMSPKPQRQRGHKNGQGENDNISVAVPKSRVQQNKSIELTAEDISLLKKMDEPFTEDTSEI Amino acids M1 – I448 (end). Residue M232 of the fusion protein is equivalent to M1 of the native enzyme. The GST tag is located at residues 1 – 220. Protease Cleavage: PreScission (LEVLFQGP) residues 221 - 228
- Concentration:Batch dependent
- Formulation:Liquid. 50 mM Tris-HCl pH 7.5, 270 mM Sucrose, 150 mM NaCl, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35
- Shipping temperature:Dry ice
Specifications
About this item
Customers Who Bought This Also Bought