SARS-CoV-2 (COVID-19) Recombinant NSP10 Protein (from bacteria)
Catalog #: 77134-114
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:43740578
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:Bacteria
- Species:SARS-CoV-2
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:SARS-CoV2-NSP10|SARS-CoV-2 NSP10|NSP10 SARS-CoV-2
- Protein/peptide name:ORF1ab
- Purity:Amylose Resin (0.6)
- Molecular weight:Mono-Isotopic Mass: 59, 810.55 daltons Average Mass: 59, 848.71 daltons
- Sequence:MKIEEGKLVIWINGDKGYNGLAEVGKKFEKDTGIKVTVEHPDKLEEKFPQVAATGDGPDIIFWAHDRFGGYAQSGLLAEITPDKAFQDKLYPFTWDAVRYNGKLIAYPIAVEALSLIYNKDLLPNPPKTWEEIPALDKELKAKGKSALMFNLQEPYFTWPLIAADGGYAFKYENGKYDIKDVGVDNAGAKAGLTFLVDLIKNKHMNADTDYSIAEAAFNKGETAMTINGPWAWSNIDTSKVNYGVTVLPTFKGQPSKPFVGVLSAGINAASPNKELAKEFLENYLLTDEGLEAVNKDKPLGAVALKSYEEELVKDPRIAATMENAQKGEIMPNIPQMSAFWYAVRTAVINAASGRQTVDEALKDAQTNSSSNNNNNNNNNNLGDDDDKVPEFLEVLFQGPLGSAGNATEVPANSTVLSFCAFAVDAAKAYKDYLASGGQPITNCVKMLCTHTGTGQAITVTPEANMDQESFGGASCCLYCRCHIDHPNPKGFCDLKGKYVQIPTTCANDPVGFTLKNTVCTVCGMWKGYGCSCDQLREPMLQ Amino acids A1 – Q139 (end). Residue A404 of the fusion protein is equivalent to A1 of the native enzyme. The MBP tag is located at residues 1 – 367. Protease Cleavage: PreScission (LEVLFQGP) residues 393 – 400
- Concentration:Batch dependent
- Formulation:Liquid. 50 mM Tris-HCl pH 7.5, 270 mM Sucrose, 150 mM NaCl, 0.1 mM EGTA, 0.1% 2-mercaptoethanol, 0.03% Brij-35
- Shipping temperature:Dry ice
Specifications
About this item
Customers Who Bought This Also Bought