Anti-ZZZ3 Rabbit Polyclonal Antibody
Catalog #: 76754-532
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Zinc finger, ZZ-type containing 3
- Clonality:Polyclonal
- Gene ID:26009
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Antigen symbol:ZZZ3
- Conjugation:Unconjugated
- ImmunoFluorescence:Yes
- Size:50 µl
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ATAC1|DKFZp313N0119|ZZZ3|DKFZp564I052|FLJ10362
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:Blue ice
- Immunogen:Recombinant protein of human ZZZ3.
- Purification:Affinity purification
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ZZZ3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Customers Who Bought This Also Bought