Anti-CLSTN1 Rabbit Polyclonal Antibody
Catalog # 76761-274
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Calsyntenin-1
- Antigen symbol:CLSTN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Size:50 µl
- Environmentally Preferable:
- Form:Liquid
- Gene ID:22883
- Antigen synonyms:Soluble Alc-alpha|CTF1-alpha|SAlc-alpha|KIAA0911|Calsyntenin-1|Non-classical cadherin XB31alpha|CLSTN1|CS1|Alzheimer-related cadherin-like protein|Alcadein-alpha|C-terminal fragment 1-alpha|Alc-alpha
- Modification:None
- Storage buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage temperature:Store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 29-350 of human CLSTN1 (NP_055759.3).
- Purification:Affinity purification
- Cat. no.:76761-274
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CLSTN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat