Anti-ZAP70 Mouse Monoclonal Antibody [clone: [SBZAP]] (AP (Alkaline Phosphatase))
Catalog #: 77120-876
Supplier: Prosci
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Tyrosine-protein kinase ZAP-70
- Clonality:Monoclonal
- Clone:[SBZAP]
- Gene ID:7535
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Antigen symbol:ZAP70
- Conjugation:AP (Alkaline Phosphatase)
- Flow cytometry:Yes
- Size:1 ml
- Form:Liquid
- Antigen synonyms:Tyrosine-protein kinase ZAP-70|Syk-related tyrosine kinase|zeta-chain-associated protein kinase 70
- Modification:None
- Storage buffer:Supplied in a stock solution of 50 mM Tris/1 mM MgCl2/50% glycerol, pH 8.0, containing NaN3 as a preservative.
- Storage temperature:Store vial at 2...8 °C.
- Concentration:Batch dependent
- Shipping temperature:Blue ice
- Immunogen:Human ZAP-70 (a.a. 280-309).
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ZAP70
Clonality: Monoclonal
Clone: [SBZAP]
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Customers Who Bought This Also Bought