Anti-Mouse Monoclonal Antibody [clone: [CIa]] (PE (Phycoerythrin))
77132-648
: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:[CIa]
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:lgM
- Reactivity:Dove,Quail,Chicken,Pigeon,Duck
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:MHC Class II
- Modification:None
- Storage buffer:Supplied in PBS/NaN3 and a stabilizing agent.
- Storage temperature:Store vial at 2...8 °C.
- Concentration:0.1 mg/ml
- Shipping temperature:20
- Immunogen:Splenic leukocytes from an outbred nine-week-old chicken.
- Cat. no.:77132-648
- Packaging:Vial
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: [CIa]
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: lgM
Reactivity: Chicken, Quail, Dove, Pigeon, Duck