Anti-CD1 Mouse Monoclonal Antibody [clone: [76-7-4]]
Catalog #: 77124-820
Supplier: Prosci
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CD1 protein
- Clonality:Monoclonal
- Clone:[76-7-4]
- Gene ID:396800
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Pig
- Antigen symbol:CD1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Size:0.5 mg
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:CD1.1|CD1c
- Modification:None
- Storage buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added.
- Storage temperature:Store vial at 2...8 °C.
- Concentration:0.5 mg/ml
- Shipping temperature:Blue ice
- Immunogen:Fresh dd miniature swine thymocytes.
- Tested applications:Proximity Ligation Assay
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CD1
Clonality: Monoclonal
Clone: [76-7-4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Pig
Customers Who Bought This Also Bought