To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Clonality:Polyclonal
- Host:Goat
- Isotype:IgG
- Reactivity:Dog,Rat,Mouse,Hamster,Pig,Rabbit,Horse,Tree shrew,Rhesus monkey,Chinchilla,Chicken,Human,Bovine,Guinea pig
- Conjugation:Unconjugated
- ELISA:Yes
- Size:0.2 mg
- Form:Liquid
- Modification:None
- Storage buffer:Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added.
- Storage temperature:Store vial at 2...8 °C.
- Concentration:0.4 mg/ml
- Shipping temperature:Blue ice
- Immunogen:Type III collagen.
- Purification:Purified by affinity chromatography on type III collagen covalently linked to agarose; cross-adsorbed against Collagen types I, II, IV, V, and VI for minimal reactivity.
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Dog, Chicken, Chinchilla, Horse, Guinea Pig, Hamster, Pig, Rabbit, Rhesus Monkey, Tree Shrew