Anti-SRCAP Goat Polyclonal Antibody
Catalog # 77127-710
Supplier: Prosci
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Snf2-related CREBBP activator protein
- Antigen symbol:SRCAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Size:0.1 mg
- Environmentally Preferable:
- Form:Liquid
- Gene ID:10847
- Antigen synonyms:FLJ44499|KIAA0309|helicase SRCAP|domino homolog 1|EAF1|Swi2/Snf2-related ATPase homolog|SWR1|domino homolog 2|SRCAP|Snf2-related CREBBP activator protein|DOMO1|OTTHUMP00000208671|Snf2-related CBP activator protein
- Modification:None
- Storage buffer:Supplied at 0.5 mg/ml in Tris saline, 0.02% sodium azide, pH7.3 with 0.5% bovine serum albumin.
Aliquot and store at -20°C. Minimize freezing and thawing. - Storage temperature:Aliquot and store at −20 °C. Minimize freezing and thawing.
- Concentration:500 µg/ml
- Shipping temperature:20
- Immunogen:The immunogen for this antibody is: C-QEGNDAEAQRREIE.
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. no.:77127-710
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SRCAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human