VWR® Aloe-Coated Nitrile Gloves
Supplier: VWR International
Certificates
About this item
The innovative aloe-coated nitrile glove is designed to combine the durability of nitrile with the soothing properties of aloe vera. The aloe coating provides a moisturizing and nourishing experience, keeping hands hydrated and preventing dryness and irritation.
- Aloe-coated
- 3.5 mil finger thickness
- Powder-free
- 9.5" length
- Food contact compliant
- High comfort and flexibility
- Textured fingers
Perfect for exceptional grip and tactile sensitivity, ensuring optimal performance in various industries such as healthcare, laboratory, food handling, and more.
Packaging: Gloves are packaged 200 per box, 10 boxes per case.
Specifications
- Catalog No:
- 76518-172
- 76518-176
- 76518-174
- 76518-180
- 76518-178
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- EN 455
- EN 455
- EN 455
- EN 455
- EN 455
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Food contact approved:
- Yes
- Yes
- Yes
- Yes
- Yes
- ASTM No.:
- ASTM D6319 Compliance
- ASTM D6319 Compliance
- ASTM D6319 Compliance
- ASTM D6319 Compliance
- ASTM D6319 Compliance