EZBio®pure Titanium Round Single-Use Cap Assembly with GL 45 VersaCap®
Supplier: Foxx Life Sciences
New Product
Certificates
About this item
EZBio®pure TR Single-use cap assemblies allow the customer to select individual components suited for their project needs. Assemblies take advantage of the patented VersaCap® technology, which means there is no machining necessary to produce the product. Perfect for single-use applications and to phase out costly stainless-steel apparatuses that are expensive to clean and validate.
- New ergonomic bottle shape with molded finger grips
- GL 45 VersaCap® made of polypropylene
- EZBio®pure TR Media bottles available in either PS or PETG materials
- Manufactured in a Class 100000 cleanroom
- Manufactured in an ISO 13485 Facility in North America
EZBio®pure Titanium Round (TR) Single-Use (SU) cap assembly with EZBio® tubing, plug, dip tube, and GL 45 VersaCap®. The term 'pure' refers to the type of resin used to manufacture the bottle. These assemblies are double-bagged, and supplied sterile.
Certifications: UPS Class VI resins.
Customers Who Bought This Also Bought