Influenza A (Swine H1N1) Neuraminidase (A/California/14/2009) Peptide
Catalog #: 77133-660
Supplier: Prosci
New Product
CAS Number:
Specifications
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Influenza A H1N1
- Conjugation:Unconjugated
- Size:0.05 mg
- Storage conditions:Store (up to 1 year) at −20 °C. Avoid multiple freeze/thaw cycles.
- Protein/peptide name:Neuraminidase (A/California/14/2009)
- Concentration:0.2 mg/ml
- Formulation:Liquid. PBS, pH 7.2 containing 0.1% BSA.
- Shipping temperature:Blue ice
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought