COX4 Peptide
Catalog #: 77133-032
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:84701
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:COXIV-2|dJ857M17.2|Cytochrome c oxidase subunit IV isoform 2|mitochondrial|Cytochrome c oxidase subunit 4 isoform 2|COX IV-2|COX4|COX4L2|COX4-2|COX4B
- Protein/peptide name:COX4I2
- Formulation:Lyophilized. COX4 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room Temperature
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought