CDH13/H cadherin Peptide
Catalog #: 77134-458
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:1012
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:CDH13|CDHH|H-cadherin|heart cadherin|T-cadherin|P105|cadherin-13|H-cadherin (heart)|T-cad|cadherin 13
- Protein/peptide name:CDH13/H cadherin
- Formulation:Lyophilized. CDH13 / H cadherin peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought