NEUROD2 Peptide
Catalog #: 77124-558
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:4761
- Packaging:Vial
- Protein/peptide type:Synthetic
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:NDRF|neurogenic differentiation factor 2|neurogenic basic-helix-loop-helix protein|neurogenic differentiation 2|NEUROD2|class A basic helix-loop-helix protein 1|MGC26304|neuroD-related factor|bHLHa1
- Protein/peptide name:NEUROD2
- Formulation:Lyophilized. NEUROD2 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77124-558
Specifications
About this item
Customers Who Bought This Also Bought