ANTXR2/CMG2 Peptide
Catalog # 77129-254
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/peptide type:Synthetic
- Source:
- Species:HumanRatMouse
- Size:0.1 mg
- Environmentally Preferable:
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Gene ID:118429
- Protein synonyms:MGC45856|ANTXR2|JHF|capillary morphogenesis protein 2|MGC111533|FLJ31074|anthrax toxin receptor 2|CMG-2|CMG2|ISH
- Protein/peptide name:ANTXR2/CMG2
- Formulation:Lyophilized. ANTXR2 / CMG2 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:RT
- Tested applications:Blocking
- Cat. no.:77129-254