MAP3K6 Peptide
Catalog #: 77123-014
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:9064
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Rat
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:MGC20114|ASK2|mitogen-activated protein kinase kinase kinase 6|MAPKKK6|OTTHUMP00000005063|MAP3K6|apoptosis signal-regulating kinase 2|MGC125653
- Protein/peptide name:MAP3K6
- Formulation:Lyophilized. MAP3K6 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77123-014
Specifications
About this item
Customers Who Bought This Also Bought