NCKAP1 Peptide
Catalog #: 77124-166
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:10787
- Packaging:Vial
- Protein/peptide type:Synthetic
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:KIAA0587|NAP 1|NAP1|Nck-associated protein 1|Membrane-associated protein HEM-2|HEM2|NAP125|p125Nap1
- Protein/peptide name:NCKAP1
- Formulation:Lyophilized. NCKAP1 Peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
- Cat. no.:77124-166
Specifications
About this item
Customers Who Bought This Also Bought