IL12RB1 Peptide
Catalog #: 77130-128
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:3594
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:CD212|IL12RB|IL-12R-BETA1|IL-12 receptor beta component|IL12R|Interleukin-12 receptor subunit beta-1|IL-12 receptor subunit beta-1
- Protein/peptide name:IL12RB1
- Formulation:Lyophilized. IL12RB1 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought