Itih4 Peptide
Catalog #: 77131-826
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:16427
- Packaging:Vial
- Protein/peptide type:Synthetic
- Species:Rat
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Store at −20 °C. When reconstituted minimize freezing and thawing.
- Protein synonyms:heavy chain 4|ITI-HC4|Itih-4|ITI heavy chain H4|Inter alpha-trypsin inhibitor
- Protein/peptide name:Itih4
- Formulation:Lyophilized. Itih4 peptide is supplied as 100 µg of dried peptide. When the peptide is reconstituted in 200 µl water, the concentration will be 0.5 mg/ml.
- Shipping temperature:Room temperature
- Tested applications:Blocking
Specifications
About this item
Customers Who Bought This Also Bought