Human Recombinant PVRIG Protein (from HEK293 cells)
Catalog #: 77125-148
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:79037
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:PVRIG|CD112R|MGC104322|CD112 receptor|C7orf15MGC138295|MGC138297|MGC2463|C7orf15
- Protein/peptide name:PVRIG
- Purity:>90% as determined by SDS-PAGE
- Molecular weight:15.6 kDa
- Sequence:Thr 41 - Asp 171
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping temperature:Room temperature
- Tested applications:ELISA,WB
Specifications
About this item
Customers Who Bought This Also Bought