Cynomolgus monkey Recombinant Serum Albumin Protein (from HEK293 cells)
Catalog #: 77126-522
Supplier: Prosci
New Product
CAS Number:
Specifications
- Gene ID:102130757
- Packaging:Vial
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Cynomolgus monkey
- Conjugation:Unconjugated
- Size:0.1 mg
- Storage conditions:Lyophilized protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Protein synonyms:Serum albumin|Alb|ALB
- Protein/peptide name:ALB
- Purity:>95% as determined by SDS-PAGE
- Molecular weight:67.9 kDa
- Sequence:Asp 25 - Ala 608
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping temperature:Room temperature
- Tested applications:WB
Specifications
About this item
Customers Who Bought This Also Bought